Anti-Listeria Chicken Polyclonal Antibody
Catalog # PRSIXW-7705
Supplier: ProSci Inc.
Specifications
- Antigen name:Listeria
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Chicken
- Reactivity:Firefly
- Western blot:Yes
- Form:Liquid
- Concentration:1 mg/mL
- Immunogen:1-550
- Tested applications:Immunoaffinity Purified
- Size:50 µg
- Pk:50 µG
Specifications
About this item
FUNCTION: Produces green light with a wavelength of 562 nM.
CATALYTIC ACTIVITY: Photinus luciferin + O2 + ATP = oxidized Photinus luciferin + CO2 + AMP + diphosphate + light.
SIMILARITY: Belongs to the ATP-dependent AMP-binding enzyme family.
COFACTOR: Magnesium.
SUBCELLULAR LOCATION: Peroxisomal.
Luciferin 4-monooxygenase antibody can be used for the detection of Luciferin 4-monooxygenase by Western Blot.
Phosphate-Buffered Saline. No preservatives added.
Phosphate-Buffered Saline. No preservatives added.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: Listeria
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Firefly