Anti-GRIN2A Rabbit Polyclonal Antibody
PRSI50-121
- Antigen name:Glutamate Ionotropic Receptor NMDA Type Subunit 2A
- Antigen symbol:GRIN2A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:NR2A|LKS|glutamate ionotropic receptor NMDA type subunit 2A|EPND|GluN2A|NMDAR2A|FESD
- Immunogen:Phosphopeptide corresponding to amino acid residues surrounding the phospho-Tyr1325 of rat NMDA NR2A.
- Size:100 µl
- Pk:100 µl
Affinity Purified
Affinity Purified
100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
Type:
Antigen: Grin2a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat