Anti-GRIN2C Rabbit Polyclonal Antibody
Catalog # PRSI50-113
Supplier: ProSci Inc.
Specifications
- Antigen name:Glutamate Ionotropic Receptor NMDA Type Subunit 2C
- Antigen symbol:GRIN2C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:NMDAR2C|GluN2C|NR2C
- Immunogen:Synthetic phospho-peptide surrounding the phospho Ser1096 of the NR2C-subunit of rat NMDA receptor.
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Affinity Purified
Affinity Purified
100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
Type:
Antigen: Grin2c
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat