- Antigen name:Oxytocin Receptor
- Antigen symbol:OXTR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Form:Liquid
- Antigen synonyms:OTR
- Immunogen:Oxytocin Receptor antibody was raised against a peptide located in the C-Terminal cytoplasmic domain of Oxytocin Receptor (Human).
- Tested applications:Immunoaffinity Chromatography
- Size:50 µg
- Pk:50 µG
The oxytocin receptor belongs to the vasopressin/oxytocin family of GPCRs because it binds similar cyclic nonapeptide hormones. The oxytocin receptor functions primarily during labor and lactation. There is a dramatic increase in the number of oxytocin receptors just before the onset of labor causing the uterine muscles to become exceedingly sensitive to oxytocin. The activated oxytocin receptor causes an inward membrane current that results in smooth muscle contraction during labor and milk ejection during lactation. This receptor also plays a role in natriuresis, kaliuresis and may have an antistress effect.
Oxytocin Receptor antibody can be used in immunohistochemistry starting at 8 μg/mL.
PBS, 0.1% sodium azide.
PBS, 0.1% sodium azide.
Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: OXTR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human