Anti-SCOC Goat Polyclonal Antibody
PRSI42-426
- Antigen symbol:SCOC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Concentration:500 ug/mL
- Immunogen:SCOC antibody was raised against a 15 amino acid synthetic peptide near the C or N-Terminus of SCOC.
- Size:100 µg
- Pk:100 µG
SCOC antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
SCOC antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SCOC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human