Anti-VEGFA Rabbit Polyclonal Antibody (Biotin) [clone: ]
Catalog # PRSI38-207
Supplier: ProSci Inc.
Specifications
- Antigen name:Vascular Endothelial Growth Factor A
- Antigen symbol:VEGFA
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Reactivity:Rat
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:VEGF|VEGF-A,|Vascular permeability factor|Vascular Endothelial Growth Factor|Vascular endothelial growth factor A
- Immunogen:Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant Rat VEGF (Rat Vascular Endothelial Growth Factor). Rat VEGF specific antibody was purified by affinity chromatography and then biotinylated.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
The reconstituted antibody can temporarily be stored at 2-8°C. Long term storage at -20°C or below. Avoid repeated freeze-thaw cycles.
Type:
Antigen: VEGFA
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat