Anti-Klebsiella Rabbit Polyclonal Antibody (Biotin)
Catalog # PRSI35-296
Supplier: ProSci Inc.
Specifications
- Antigen name:Klebsiella
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Rabbit
- Reactivity:Bacteria
- Western blot:Yes
- Form:Liquid
- Immunogen:Klebsiella pneumoniae
- Size:1 mL
- Pk:1 mL
Specifications
About this item
Purified IgG fraction of antiserum covalently coupled with the N-Hydroxysuccinimide ester of biotin under mild conditions to give a high degree of substitution.
Klebsiella sp. antibody is in a phosphate saline buffer (0.01M, pH 7.2) containing 0.1% sodium azide preservative. No stabilizing proteins have been added.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: Klebsiella
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bacteria