Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-ST8SIA2 Rabbit Polyclonal Antibody
  PRSI30-217
undefined
Anti-ST8SIA2 Rabbit Polyclonal Antibody
  PRSI30-217

 

  • Antigen name:
    ST8 Alpha-N-Acetyl-Neuraminide Alpha-2,8-Sialyltransferase 2
  • Antigen symbol:
    ST8SIA2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Dog,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antigen synonyms:
    HsT19690|SIAT8B|ST8SIA-II|STX
  • Immunogen:
    A synthetic peptide corresponding a region of human ST8SIA2.
  • Size:
    50 µg
  • Pk:
    50 µG

 

 

ST8SIA2 is a type II membrane protein that is thought to catalyze the transfer of sialic acid from CMP-sialic acid to N-linked oligosaccharides and glycoproteins. ST8SIA2 may be found in the Golgi apparatus and may be involved in the production of polysialic acid, a modulator of the adhesive properties of neural cell adhesion molecule (NCAM1). This protein is a member of glycosyltransferase family 29The protein encoded by this gene is a type II membrane protein that is thought to catalyze the transfer of sialic acid from CMP-sialic acid to N-linked oligosaccharides and glycoproteins. The encoded protein may be found in the Golgi apparatus and may be involved in the production of polysialic acid, a modulator of the adhesive properties of neural cell adhesion molecule (NCAM1). This protein is a member of glycosyltransferase family 29.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ST8SIA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human, Mouse, Rat