Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-SLC26A5 Rabbit Polyclonal Antibody
Catalog # PRSI29-962
Supplier:  ProSci Inc.
undefined
Anti-SLC26A5 Rabbit Polyclonal Antibody
Catalog # PRSI29-962
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Solute Carrier Family 26 Member A5
  • Antigen symbol:
    SLC26A5
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Dog,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Immunogen:
    A synthetic peptide corresponding a region of human SLC26A5.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

SLC26A5 is a member of the SLC26A/SulP transporter family. SLC26A5 is specifically expressed in outer hair cells (OHCs) of the cochlea and is essential in auditory processing. Intracellular anions are thought to act as extrinsic voltage sensors, which bind to this protein and trigger the conformational changes required for rapid length changes in OHCs. Mutations in its gene have been associated with non-syndromic hearing loss.This gene is a member of the SLC26A/SulP transporter family. It encodes a protein that is specifically expressed in outer hair cells (OHCs) of the cochlea and is essential in auditory processing. Intracellular anions are thought to act as extrinsic voltage sensors, which bind to this protein and trigger the conformational changes required for rapid length changes in OHCs. Mutations in this gene have been associated with non-syndromic hearing loss. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SLC26A5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human, Mouse, Rat