Specifications
- Antigen name:Solute Carrier Family 22 Member A3
- Antigen symbol:SLC22A3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:EMT|EMTH|OCT3|extraneuronal monoamine transporter member 3|organic cation transporter 3|solute carrier family 22|solute carrier family 22 member 3|EMT organic cation transporter 3|extraneuronal monoamine transporter
- Immunogen:A synthetic peptide corresponding a region of human SLC22A3.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Polyspecific organic cation transporters in the liver, kidney, intestine, and other organs are critical for elimination of many endogenous small organic cations as well as a wide array of drugs and environmental toxins. SLC22A3 contains twelve putative transmembrane domains and is a plasma integral membrane protein.Polyspecific organic cation transporters in the liver, kidney, intestine, and other organs are critical for elimination of many endogenous small organic cations as well as a wide array of drugs and environmental toxins. This gene is one of three similar cation transporter genes located in a cluster on chromosome 6. The encoded protein contains twelve putative transmembrane domains and is a plasma integral membrane protein.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SLC22A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human