Order Entry
Czech Republic
ContactUsLinkComponent
Anti-SLC17A5 Rabbit Polyclonal Antibody
Anti-SLC17A5 Rabbit Polyclonal Antibody
Catalog # PRSI29-924
Supplier:  ProSci Inc.
undefined
Anti-SLC17A5 Rabbit Polyclonal Antibody
Catalog # PRSI29-924
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Solute Carrier Family 17 Member A5
  • Antigen symbol:
    SLC17A5
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Immunogen:
    A synthetic peptide corresponding a region of human SLC17A5.
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

SLC17A5 is a membrane transporter that exports free sialic acids that have been cleaved off of cell surface lipids and proteins from lysosomes. Mutations in SLC17A5 gene cause sialic acid storage diseases, including infantile sialic acid storage disorder and and Salla disease, an adult form.This gene encodes a membrane transporter that exports free sialic acids that have been cleaved off of cell surface lipids and proteins from lysosomes. Mutations in this gene cause sialic acid storage diseases, including infantile sialic acid storage disorder and and Salla disease, an adult form. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SLC17A5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat