- Antigen name:TEA Domain Family member 3
- Antigen symbol:TEAD3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Dog,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:TEF-5|DTEF-1|TEAD5|ETFR-1|TEF5|TEAD-3
- Immunogen:A synthetic peptide corresponding a region of human TEAD3.
- Size:50 µg
- Pk:50 µG
TEAD3 is a member of the transcriptional enhancer factor (TEF) family. The family members contain the TEA/ATTS DNA-binding domain. This member is preferentially expressed in placenta and transactivates the chorionic somatomammotropin gene enhancer.This gene encodes a member of the transcriptional enhancer factor (TEF) family. The family members contain the TEA/ATTS DNA-binding domain. This member is preferentially expressed in placenta and transactivates the chorionic somatomammotropin gene enhancer. The protein is encoded through the use of a non-AUG (ATA) translation initiation codon.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: TEAD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human, Mouse, Rat