Anti-MYNN Rabbit Polyclonal Antibody
Catalog # PRSI27-798
Dodavatel: ProSci Inc.
Specifikace
- Antigen name:Myoneurin
- Antigen symbol:MYNN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Immunogen:A synthetic peptide corresponding a region of human MYNN.
- Size:100 µg
- Pk:100 µG
Specifikace
O tomto produktu
Myoneurin belongs to the BTB/POZ and zinc finger protein family. The BTB/POZ and zinc finger proteins (BTB/POZ-ZF) constitute a growing family of proteins with gene expression regulatory functions since they have been shown to be involved in both transcriptional activation and repression of various genes in a broad range of species.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: MYNN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human