Anti-INSM1 Rabbit Polyclonal Antibody
Catalog # PRSI27-664
Supplier: ProSci Inc.
Specifications
- Antigen name:Insulinoma-associated 1
- Antigen symbol:INSM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:IA1|IA-1|Zinc finger protein IA-1|Insulinoma-associated protein 1
- Immunogen:A synthetic peptide corresponding a region of human INSM1.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
The insulinoma-associated 1 (INSM1) gene is intronless and encodes a protein containing both a zinc finger DNA-binding domain and a putative prohormone domain. This gene is a sensitive marker for neuroendocrine differentiation of human lung tumors.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: INSM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human