- Antigen symbol:UNC5A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:Unc 5 homolog 1|Unc 5 homolog A|Protein unc-5 homolog A|Netrin receptor Unc5h1|Netrin receptor UNC5A|Netrin receptor UNC 5A|UNC5A_HUMAN|Unc 5a|Unc5h1|KIAA1976.|Unc5a|Unc5 a|Unc5 homolog 1|FLJ16449|Protein unc-5 homolog 1|Unc5 homolog A
- Immunogen:A synthetic peptide corresponding a region of human UNC5A.
- Size:50 µg
- Pk:50 µG
UNC5A belongs to a family of netrin-1 receptors thought to mediate the chemorepulsive effect of netrin-1 on specific axons. For more information on UNC5 proteins, see UNC5C.UNC5A belongs to a family of netrin-1 (MIM 601614) receptors thought to mediate the chemorepulsive effect of netrin-1 on specific axons. For more information on UNC5 proteins, see UNC5C (MIM 603610).
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: UNC5A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat