Order Entry
Czech Republic
ContactUsLinkComponent
Anti-GMPPA Rabbit Polyclonal Antibody
Anti-GMPPA Rabbit Polyclonal Antibody
Catalog # PRSI26-386
Supplier:  ProSci Inc.
undefined
Anti-GMPPA Rabbit Polyclonal Antibody
Catalog # PRSI26-386
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    GDP Mannose Pyrophosphorylase A
  • Antigen symbol:
    GMPPA
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antigen synonyms:
    GDP mannose pyrophosphorylase A|GTP mannose 1 phosphate guanylyltransferase alpha|Mannose 1 phosphate guanylyltransferase GDP,GMPPA_HUMAN.
  • Immunogen:
    A synthetic peptide corresponding a region of human GMPPA.
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

GMPPA is a GDP-mannose pyrophosphorylase. This enzyme catalyzes the reaction which converts mannose-1-phosphate and GTP to GDP-mannose which is involved in the production of N-linked oligosaccharides.This gene is thought to encode a GDP-mannose pyrophosphorylase. This enzyme catalyzes the reaction which converts mannose-1-phosphate and GTP to GDP-mannose which is involved in the production of N-linked oligosaccharides.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GMPPA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat