Order Entry
Czech Republic
ContactUsLinkComponent
Anti-GALNT18 Rabbit Polyclonal Antibody
Anti-GALNT18 Rabbit Polyclonal Antibody
  PRSI26-384
undefined
Anti-GALNT18 Rabbit Polyclonal Antibody
  PRSI26-384

 

  • Antigen name:
    Polypeptide N-Acetylgalactosaminyltransferase 18
  • Antigen symbol:
    GALNT18
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antigen synonyms:
    GALNT15|GalNAc-T18|GalNAc-T15|GALNTL4|GALNACT18
  • Immunogen:
    A synthetic peptide corresponding a region of human GALNTL4.
  • Size:
    50 µg
  • Pk:
    50 µG

 

 

GALNTL4 may catalyze the initial reaction in O-linked oligosaccharide biosynthesis, the transfer of an N-acetyl-D-galactosamine residue to a serine or threonine residue on the protein receptor.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GALNT18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat