Specifications
- Název antigenu:SCAN Domain Containing 2 pseudogene
- Symbol antigenu:SCAND2P
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- ELISA:Yes
- Hostitel:Rabbit
- Reaktivita:Human
- Western blot:Yes
- Formulář:Lyophilized
- Imunogen:A synthetic peptide corresponding a region of human SCAND2.
- Rozměry protilátky:50 µg
- Bal.:50 µG
Specifications
About this item
The SCAN domain is a highly conserved, leucine-rich motif of approximately 60 aa originally found within a subfamily of zinc finger proteins. Functional studies have established that the SCAN box is a protein interaction domain that mediates both hetero- and homoprotein associations, and maybe involved in regulation of transcriptional activity.
SCAND2 antibody can be used for detection of SCAND2 by ELISA at 1:1562500. SCAND2 antibody can be used for detection of SCAND2 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SCAND2P
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human