Anti-ZFAND3 Rabbit Polyclonal Antibody
PRSI25-208
- Antigen name:Zinc Finger, AN1-type Domain 3
- Antigen symbol:ZFAND3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Immunogen:A synthetic peptide corresponding a region of human ZFAND3.
- Size:50 µg
- Pk:50 µG
ZFAND3 contains 1 A20-type zinc finger and 1 AN1-type zinc finger. The exact function of ZFAND3 remains unknown.
ZFAND3 antibody can be used for detection of ZFAND3 by ELISA at 1:312500. ZFAND3 antibody can be used for detection of ZFAND3 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ZFAND3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human