Order Entry
Czech Republic
ContactUsLinkComponent
Anti-OR13C5 Rabbit Polyclonal Antibody
Anti-OR13C5 Rabbit Polyclonal Antibody
Catalog # PRSI25-017
Supplier:  ProSci Inc.
undefined
Anti-OR13C5 Rabbit Polyclonal Antibody
Catalog # PRSI25-017
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Olfactory Receptor, Family 13, Subfamily C Member 5
  • Antigen symbol:
    OR13C5
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Immunogen:
    A synthetic peptide corresponding a region of human OR13C5.
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.

OR13C5 antibody can be used for detection of OR13C5 by ELISA at 1:312500. OR13C5 antibody can be used for detection of OR13C5 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: OR13C5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human