Anti-KPNA5 Rabbit Polyclonal Antibody
Catalog # PRSI5989
Supplier: ProSci Inc.
Specifications
- Antigen name:Karyopherin Alpha 5
- Antigen symbol:KPNA5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Antigen synonyms:IPOA6|SRP6|KPNA5|Karyopherin subunit alpha-5|Importin subunit alpha-6
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:KPNA5 antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human KPNA5.
- Tested applications:E, IHC
- Purification:Affinity chromatography purified via peptide column
- Size:100 µg
- Pk:100 µG
Specifications
About this item
KPNA5 Antibody: Karyopherin, a cytosolic and heterodimeric protein complex consisting of alpha and beta subunits, is responsible for targeting proteins with nuclear localization signals to the nuclear pore complex (NPC) by an energy requiring, Ran-dependent mechanism. The alpha subunit and imported substrate enter the nucleus and accumulate in the nucleoplasm, while the beta subunit accumulates at the NPC. KPNA5 belongs to a subfamily within the KPNA family that also includes KPNA4 and 6 and is thought to be involved in NLS-dependent protein import into the nucleus.
Type:
Antigen: KPNA5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human