Specifications
- Antigen name:Phosphatidic Acid Phosphatase Type 2 Domain-Containing Protein 1B
- Antigen symbol:PPAPDC1B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Antigen synonyms:HTPAP|Phosphatidate phosphatase PPAPDC1B|PPAPDC1B|DPPL1|Phosphatidic acid phosphatase type 2 domain-containing protein 1B
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:PPAPDC1B antibody was raised against a 14 amino acid synthetic peptide near the amino terminus of human PPAPDC1B.
- Tested applications:E, WB
- Purification:Affinity chromatography purified via peptide column
- Size:100 µg
- Pk:100 µG
Specifications
About this item
PPAPDC1B Antibody: Phosphatidate phosphatase (PAP) plays important role in lipid-signaling metabolism in eukaryotic cells. Two distinct types of PAP (PAP1 and PAP2) activity have been distinguished by their subcellular localization and differential sensitivity to N-ethylmaleimide (NEM) and Mg2+. A yeast diacylglycerol pyrophosphate (DGPP) phosphatase (DPP1) and mammalian DGPP phosphatase (PAP2) have been identified as Mg2+-independent and NEM-insensitive membrane-associated. PPAPDC1A (also known as DPPL2) and PPAPDC1B (DPPL1) form a novel type of Mg2+-independent and NEM-sensitive mammalian phosphatidate phosphatase showing broad substrate specificity. Knockdown experiments indicated that this protein is involved with multiple cell signaling pathways, including the JAK-Stat3, MAP kinase, and PKC pathways. PPAPDC1B may also potentiate the estrogen receptor pathway by down-regulating DUSP22.
Type:
Antigen: PPAPDC1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse