Specifications
- Antigen name:Mitogen-Activated Protein Kinase Associated Protein 1
- Antigen symbol:MAPKAP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Antigen synonyms:TORC2 subunit MAPKAP1|MAPKAP1|SIN1|MIP1|Mitogen-activated protein kinase 2-associated protein 1|JC310|SIN1b|Target of rapamycin complex 2 subunit MAPKAP1|SIN1g
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:MAPKAP1 antibody was raised against a 18 amino acid synthetic peptide from near the center of human MAPKAP1.
- Tested applications:E, WB, IHC
- Purification:Affinity chromatography purified via peptide column
- Size:100 µg
- Pk:100 µG
Specifications
About this item
MAPKAP1 Antibody: MAPKAP1 was initially identified as the human homolog of S. pombe SIN1. Recent evidence has shown that it identical to Mip1, a protein that interacts with MEKK2, a member of the mitogen-activated protein kinase (MAPK) intracellular signaling network. MAPKAP1 is thought to prevent MEKK2 activation and dimerization by forming a complex with the inactive and non-phosphorylated MEKK2, thereby blocking the JNK1/2, ERK1/2, p38 and ERK5 MAPKs. MAPKAP1 has also been shown to play a role in the TOR signaling process, a pathway that is involved in controlling cell growth and proliferation in response to environmental cues such as nutrients, growth factors and hormones. Experiments showed that MAPKAP1 helped to maintain the TOR/rictor assembly but not the TOR/RAPTOR complex, which allowed specific phosphorylation of Akt, a kinase that is believed to couple the growth factor-PI3K signaling pathway to the nutrient-regulated TOR signaling pathway. Multiple alternatively spliced isoforms of MAPKAP1 have been identified.
Type:
Antigen: MAPKAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat