- Antigen name:Lysosomal-associated membrane protein 1
- Antigen symbol:LAMP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Antigen synonyms:LGP120|CD107a|CD107 antigen-like family member A|Lysosome-associated membrane glycoprotein 1|LAMP-1|LAMPA
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:LAMP-1 antibody was raised against a 15 amino acid synthetic peptide from near the center of human LAMP-1.
- Tested applications:E, WB, IHC, IF
- Purification:Affinity chromatography purified via peptide column
- Size:100 µg
- Pk:100 µG
LAMP-1 Antibody: Autophagy, the process of bulk degradation of cellular proteins through an autophagosomic-lysosomal pathway is important for normal growth control and may be defective in tumor cells. It is involved in the preservation of cellular nutrients under starvation conditions as well as the normal turnover of cytosolic components and is negatively regulated by TOR (Target of rapamycin). A protein recently found to be involved in autophagy, LAMP-2, is a highly glycosylated protein associated with the lysosome. LAMP-1 shares much homology to LAMP-2 and is thought to have overlapping functions. Mice lacking LAMP-1 had very minor defects compared to those deficient in LAMP-2 expression. However, the loss of both proteins resulted in embryonic lethality, suggesting that each protein possesses some unique and necessary functions.
Type:
Antigen: LAMP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat