Specifications
- Antigen name:Tuberous sclerosis 2 protein
- Antigen symbol:TSC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Antigen synonyms:TSC4|LAM|TSC2|Tuberous sclerosis 2 protein|Tuberin
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:TSC2 antibody was raised against a 15 amino acid synthetic peptide from near the carboxy terminus of human TSC2.
- Tested applications:E, WB
- Purification:Affinity chromatography purified via peptide column
- Size:100 µg
- Pk:100 µG
Specifications
About this item
TSC2 Antibody: Tuberous sclerosis complex (TSC) is an autosomal dominant tumor syndrome caused by mutations in either of the TSC1 or TSC2 tumor suppressor genes. The products of these genes form a protein complex that indirectly decreases the signaling of the mammalian Target of Rapamycin (TOR), an evolutionarily conserved serine/threonine kinase that regulates cell growth and cell cycle through its ability to integrate signals from nutrient levels and growth factors. TOR activity is stimulated by Rheb, a member of the Ras superfamily of G-proteins, when the GTP/GDP ratio bound to Rheb is high. Immunoprecipitated TSC1/TSC2 has been shown to stimulate Rheb GTPase activity in vitro, suggesting that the TSC1/TSC2 complex decreases the ability of Rheb to stimulate TOR activity. This is supported by experiments showing that overexpression of TSC1 and TSC2 results in a significant decrease in the GTP/GDP ratio bound to Rheb and the inhibition of cell growth. At least three isoforms of TSC2 exist.
Type:
Antigen: TSC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse