Specifications
- Antibody type:Primary
- Antigen name:Programmed Death-ligand 1
- Antigen symbol:PD-L1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Antigen synonyms:PDL1|PD-L1|B7H1|CD274 molecule|B7-H|PDCD1L1|PDCD1LG1
- Storage buffer:PDL-1 Antibody is supplied in PBS containing 0.02% sodium azide.
- Tested applications:E, WB, IHC, IF
- Size:100 µl
- Pk:100 µG
Specifications
About this item
PDL-1 Antibody: Cell-mediated immune responses are initiated by T lymphocytes that are themselves stimulated by cognate peptides bound to MHC molecules on antigen-presenting cells (APC). T-cell activation is generally self-limited as activated T cells express receptors such as PD-1 (also known as PDCD-1) that mediate inhibitory signals from the APC. PD-1 can bind two different but related ligands, PDL-1 and PDL-2. PDL-1 is a B7-related protein that inhibits cell-mediated immune responses by reducing the secretion of IL-2 and IL-10 from memory T cells. This suggests that PDL-1 may be useful in reducing allogenic CD4+ memory T-cell responses to endothelial cells, thereby reducing the likelihood of host immune responses to allografts. At least two isoforms of PDL-1 are known to exist; this antibody is specific to the larger isoform. PDL-1 antibody has no cross-reactivity to PDL-2.
PDL-1 antibody was raised against a 17 amino acid synthetic peptide from near the center of human PDL-1.
Type: Primary
Antigen: PD-L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat