Order Entry
Czech Republic
ContactUsLinkComponent
 
Cefaclor, Reference Standards
 :  USP
 
Cefaclor, Reference Standards
 :  USP

 

  • CAS No.:
    70356-03-5
  • Formula:
    C₁₅H₁₆ClN₃O₅S
  • Description:
    Cefaclor

 

Frequently Bought Together