Order Entry
Czech Republic
ContactUsLinkComponent
 
Venlafaxine Related Compounds, Reference Standard
 :  USP
 
Venlafaxine Related Compounds, Reference Standard
 :  USP

 

  • CAS No.:
    93413-90-2
  • Synonym(s):
    1-(1-(4-Methoxyphenyl)-2-(methylamino)ethyl)cyclohexanol hydrochloride
  • Formula:
    C₁₇H₂₇NO₂
  • Description:
    Venlafaxine Related Compound A
  • UN No.:
    3077

 

Frequently Bought Together