Emergency stop safety covers
Supplier: Brady
Certificates
About this item
These steel devices guard against unauthorised operation or accidental activation.
- Can be permanently installed over push/pull or twist-release emergency stops
- Durable metal bar locks button in the depressed (OFF) position when lock shackle is inserted
- For use with standard IEC 22,50 mm or NEMA 30,5 mm buttons
Frequently Bought Together