Anti-VEGFB Rabbit Polyclonal Antibody
BSBTA04494-3
- Druh protilátky:Primární
- Symbol antigenu:VEGFB
- Klonovalita:Polyclonal
- Hostitel:Rabbit
- Reaktivita:Human,Rat,Mouse
- Rozměry protilátky:100 µg
- Bal.:100 µG
Rabbit IgG polyclonal antibody for VEGFB detection. Tested with WB, Direct ELISA in Human;Mouse;Rat.
Type: Primary
Antigen: VEGFB
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat