Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-IL27 Rabbit Polyclonal Antibody
  BSBTA00857-1
Anti-IL27 Rabbit Polyclonal Antibody
  BSBTA00857-1

 

  • Antibody type:
    Primary
  • Antigen symbol:
    IL27
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Rabbit IgG polyclonal antibody for IL27 detection. Tested with WB, ELISA(Cap) in Human.

Type: Primary
Antigen: IL27
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human