Anti-IL17C Rabbit Polyclonal Antibody
BSBTA00164-2
- Antibody type:Primary
- Antigen symbol:IL17C
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Mouse
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for IL17C detection. Tested with WB, ELISA(Cap) in Human;Mouse.
Type: Primary
Antigen: IL17C
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse