Order Entry
Czech Republic
ContactUsLinkComponent
Anti-COL18A1 Rabbit Polyclonal Antibody
Anti-COL18A1 Rabbit Polyclonal Antibody
Catalog # BSBTA01302-1
Supplier:  Boster Bio
Anti-COL18A1 Rabbit Polyclonal Antibody
Catalog # BSBTA01302-1
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    COL18A1
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for COL18A1 detection. Tested with WB, ELISA(Cap) in Human;Mouse;Rat.

Type: Primary
Antigen: COL18A1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat