Anti-DCK Rabbit Polyclonal Antibody
Catalog # BSBTA01655-1
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:DCK
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for DCK detection. Tested with WB, Direct ELISA in Human;Mouse;Rat.
Type: Primary
Antigen: DCK
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat