Anti-Caspase 4 Rabbit Polyclonal Antibody
Catalog # BSBTA02941-1
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Caspase 4
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Caspase 4 detection. Tested with WB, IHC-P, Direct ELISA in Human.
Type: Primary
Antigen: Caspase 4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human