Anti-CD11c Rabbit Polyclonal Antibody
Catalog # BSBTA00357-1
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:CD11c
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Mouse
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for CD11c detection. Tested with WB, IHC-P, Direct ELISA in Human;Mouse.
Type: Primary
Antigen: CD11c
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse