Anti-GSK3 alpha Rabbit Polyclonal Antibody
BSBTA03152-2
- Antibody type:Primary
- Antigen name:GSK3 alpha
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for GSK3 alpha detection. Tested with WB in Human;Mouse;Rat.
Type: Primary
Antigen: GSK3 alpha
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat