Anti-CD8 alpha Rabbit Polyclonal Antibody
BSBTA02236-1
- Antibody type:Primary
- Antigen name:CD8 alpha
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Rat,Mouse
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for CD8 alpha detection. Tested with WB, IHC-P, FCM, Direct ELISA in Mouse;Rat.
Type: Primary
Antigen: CD8 alpha
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse;Rat