Anti-TFF3 Rabbit Polyclonal Antibody
BSBTA01738-3
- Druh protilátky:Primární
- Symbol antigenu:TFF3
- Klonovalita:Polyclonal
- Hostitel:Rabbit
- Reaktivita:Rat,Mouse
- Rozměry protilátky:100 µg
- Bal.:100 µG
Rabbit IgG polyclonal antibody for TFF3 detection. Tested with WB, IHC-P, Direct ELISA in Mouse;Rat.
Type: Primary
Antigen: TFF3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse;Rat