Zadání objednávky
Czech Republic
ContactUsLinkComponent
 
Anti-PDK2 Rabbit Polyclonal Antibody
  BSBTA03261
Anti-PDK2 Rabbit Polyclonal Antibody
  BSBTA03261

 

  • Druh protilátky:
    Primární
  • Symbol antigenu:
    PDK2
  • Klonovalita:
    Polyclonal
  • Hostitel:
    Rabbit
  • Reaktivita:
    Human,
    Rat,
    Mouse
  • Rozměry protilátky:
    100 µg
  • Bal.:
    100 µG

 

 

Rabbit IgG polyclonal antibody for PDK2 detection. Tested with WB, Direct ELISA in Human;Mouse;Rat.

Type: Primary
Antigen: PDK2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat