Anti-FAP Rabbit Polyclonal Antibody
BSBTA00422-1
- Antibody type:Primary
- Antigen symbol:FAP
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Rat,Mouse
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for FAP detection. Tested with WB, Direct ELISA in Human;Mouse;Rat.
Type: Primary
Antigen: FAP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse;Rat