Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-CASP6 Rabbit Polyclonal Antibody
  BSBTA02631
Anti-CASP6 Rabbit Polyclonal Antibody
  BSBTA02631

 

  • Antibody type:
    Primary
  • Antigen name:
    Caspase-6
  • Antigen symbol:
    CASP6
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Rabbit IgG polyclonal antibody for Caspase-6(CASP6) detection. Tested with WB in Human;Mouse;Rat.

Type: Primary
Antigen: CASP6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat