Order Entry
Czech Republic
ContactUsLinkComponent
Anti-RANBP1 Rabbit Polyclonal Antibody
Anti-RANBP1 Rabbit Polyclonal Antibody
Catalog # BSBTA04000-2
Supplier:  Boster Bio
Anti-RANBP1 Rabbit Polyclonal Antibody
Catalog # BSBTA04000-2
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Ran-specific GTPase-activating protein
  • Antigen symbol:
    RANBP1
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for Ran-specific GTPase-activating protein(RANBP1) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Type: Primary
Antigen: RANBP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat