Anti-HLA-B Rabbit Polyclonal Antibody
Catalog # BSBTA00186-1
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:MHC class I antigen
- Antigen symbol:HLA-B
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for MHC class I antigen(HLA-B) detection. Tested with WB in Human.
Type: Primary
Antigen: HLA-B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human