Anti-HDAC4 Rabbit Polyclonal Antibody
BSBTA00971-1
- Antibody type:Primary
- Antigen name:Histone deacetylase 4
- Antigen symbol:HDAC4
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for Histone deacetylase 4(HDAC4) detection. Tested with WB in Human;Mouse;Rat.
Type: Primary
Antigen: HDAC4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat