Anti-GDF15 Rabbit Polyclonal Antibody
BSBTA01583-1
- Antibody type:Primary
- Antigen name:Growth/differentiation factor 15
- Antigen symbol:GDF15
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for Growth/differentiation factor 15(GDF15) detection. Tested with WB in Human.
Type: Primary
Antigen: GDF15
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human