Order Entry
Czech Republic
ContactUsLinkComponent
Anti-AKR1B10 Rabbit Polyclonal Antibody
Anti-AKR1B10 Rabbit Polyclonal Antibody
Catalog # BSBTA02976
Supplier:  Boster Bio
Anti-AKR1B10 Rabbit Polyclonal Antibody
Catalog # BSBTA02976
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Aldo-keto reductase family 1 member B10
  • Antigen symbol:
    AKR1B10
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for Aldo-keto reductase family 1 member B10(AKR1B10) detection. Tested with WB, IHC-P in Human.

Type: Primary
Antigen: AKR1B10
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human