Order Entry
Czech Republic
ContactUsLinkComponent
Azithromycin, Reference Standards
Azithromycin, Reference Standards
Supplier:  USP
Certificates
Azithromycin, Reference Standards
Supplier:  USP

Specifications

  • CAS No.:
    117772-70-0
  • Formula:
    C₃₈H₇₂N₂O₁₂
  • Description:
    Azithromycin
  • Synonym(s):
    3'-N-demethyl-3'-N-formylazithromycin

Specifications

Frequently Bought Together