Anti-Flt3 ligand Rabbit Polyclonal Antibody
Catalog # BSBTA04987-1
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Flt3 ligand
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:
- Reactivity:Rat,Mouse
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Flt3 ligand detection. Tested with WB, Direct ELISA in Mouse;Rat.
Type: Primary
Antigen: Flt3 ligand
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat