Anti-B2M Rabbit Polyclonal Antibody
Katalogové číslo BSBTA00456-1
Dodavatel: Boster Bio
Specifikace
- Antibody type:Primary
- Antigen name:Beta-2-microglobulin
- Antigen symbol:B2M
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifikace
O tomto produktu
Rabbit IgG polyclonal antibody for Beta-2-microglobulin(B2M) detection. Tested with WB, IHC-P in Human.
Type: Primary
Antigen: B2M
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human